Learn More
Prothipendyl Hydrochloride, TRC
Product Code. 30008967
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008967 25 mg 25mg
30005815 10 mg 10mg
30006163 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30008967 Supplier Toronto Research Chemicals Supplier No. TRCP83885025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1225-65-6
Molecular Formula C16 H19 N3 S . Cl H
Molecular Weight (g/mol) 321.87
Synonym 10H-Pyrido[3,2-b][1,4]benzothiazine, 10-(3-dimethylaminopropyl)-, hydrochloride (6CI), 10H-Pyrido[3,2-b][1,4]benzothiazine-10-propanamine, N,N-dimethyl-, monohydrochloride (9CI), 10H-Pyrido[3,2-b][1,4]benzothiazine-10-propanamine, N,N-dimethyl-, hydrochloride (1:1), Prothipendyl Hydrochloride
IUPAC Name N,N-dimethyl-3-pyrido[3,2-b][1,4]benzothiazin-10-ylpropan-1-amine;hydrochloride
SMILES Cl.CN(C)CCCN1c2ccccc2Sc3cccnc13
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C16H19N3S.ClH/c1-18(2)11-6-12-19-13-7-3-4-8-14(13)20-15-9-5-10-17-16(15)19;/h3-5,7-10H,6,11-12H2,1-2H3;1H
Formula Weight 321.1066
Percent Purity >95%
Chemical Name or Material Prothipendyl Hydrochloride
Grade HPLC