missing translation for 'onlineSavingsMsg'
Learn More

Prothipendyl Hydrochloride, TRC

Product Code. 30005815
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005815 10 mg 10mg
30006163 5 mg 5mg
30008967 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005815 Supplier Toronto Research Chemicals Supplier No. TRCP83885010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1225-65-6
Molecular Formula C16 H19 N3 S . Cl H
Molecular Weight (g/mol) 321.87
Synonym 10H-Pyrido[3,2-b][1,4]benzothiazine, 10-(3-dimethylaminopropyl)-, hydrochloride (6CI), 10H-Pyrido[3,2-b][1,4]benzothiazine-10-propanamine, N,N-dimethyl-, monohydrochloride (9CI), 10H-Pyrido[3,2-b][1,4]benzothiazine-10-propanamine, N,N-dimethyl-, hydrochloride (1:1), Prothipendyl Hydrochloride
IUPAC Name N,N-dimethyl-3-pyrido[3,2-b][1,4]benzothiazin-10-ylpropan-1-amine;hydrochloride
SMILES Cl.CN(C)CCCN1c2ccccc2Sc3cccnc13

Especificaciones

Quantity 10 mg
InChI Formula InChI=1S/C16H19N3S.ClH/c1-18(2)11-6-12-19-13-7-3-4-8-14(13)20-15-9-5-10-17-16(15)19;/h3-5,7-10H,6,11-12H2,1-2H3;1H
Formula Weight 321.1066
Percent Purity >95%
Chemical Name or Material Prothipendyl Hydrochloride
Grade HPLC
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.