Learn More
Thermo Scientific™ Proteinase K, Tritirachium album limber
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Content And Storage | Keep cold |
| Quantity | 100 mg |
| Form | Lyophilized Powder |