Learn More
Novus Biologicals™ Protein A Native Protein
Shop All Bio Techne Products
Click to view available options
Unit Size:
5mL
Description
Native Protein A. The Protein A Native Protein has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
| Concentration | 6 mg/mL |
| Gene ID (Entrez) | 3919448 |