Learn More
Mutexil™ Protective Overshoe Coverings
Product Code. 11780124
Change view
Click to view available options
Product Type:
Overshoe
Protective CPE Visitor Overshoes
Sticky Overshoe
Visitor Overshoe
Color:
Blue
White
White, Blue
Size:
2X-Large
Universal
X-Large
Unit Size:
Pack of 2000
Pack of 300
Pack of 600
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Color Size Unit Size
11780124 Protective CPE Visitor Overshoes Blue Universal Pack of 2000
11770124 Protective CPE Visitor Overshoes Blue X-Large Pack of 2000
11790124 Protective CPE Visitor Overshoes White Universal Pack of 2000
15477933 Visitor Overshoe White, Blue Universal Pack of 600
11775433 Sticky Overshoe White 2X-Large Pack of 300
11765433 Overshoe Blue 2X-Large Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11780124 Supplier Mutexil™ Supplier No. 3335113
  • ONLINE EXCLUSIVE PRICE
    133.35 EUR
    105.67 EUR / Pack of 2000
    Save 27.68 EUR
    20% Off
Estimated Shipment: 07-10-2026
Choose from a range of choices and construction, from sticky, anti-slip, fluid -resistant, polypropylene and CPE variations with Mutexil™ Protective Overshoe Coverings.
  • Various sizes and colors
  • Needs-based styles
  • Per new standards EN ISO 20345 and EN ISO 20347
Specifica
Color Blue
For Use With (Application) For sanitary, food, industrial, catering and services
Material Polyethylene
Size Universal
Style CPE
Waterproof Yes
Product Type Protective CPE Visitor Overshoes
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.