Learn More
3M™ Protective Coverall 4565
Product Code. 17805666
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 20
Product Code. Size unitSize
17805666 Large Pack of 20
17855676 Small Pack of 20
17845666 Medium Pack of 20
17865666 X-Large Pack of 20
17875666 2X-Large Pack of 20
17845676 3X-Large Pack of 20
17875676 4X-Large Pack of 20
7 options
Product Code. 17805666
Brand: 3M™ 7100256144
Only null left
Add to basket
Add to basket
Protective Coverall 4565 has desirable comfort features and provides a strong barrier to certain chemicals, liquid sprays and particles.
  • Wear 3M™ Protective Coverall 4565 for an ideal barrier to hazardous particles, limited liquid splashes and sprays to Type 4
  • Fresh white coverall 4565 has recognizable red taped seams and meets a variety of standards, including chemical protective clothing EN 14126 Infective agents, EN 1073-2 Nuclear particulate resistance, EN 1149, EN ISO 13688, Type 6 Liquid Splash EN 13034 and Type 5 Dust EN ISO 13982-1, and Type 4 Liquid Spray EN 14605
  • Taped seams reduce skin exposure points through tearing and reduced risk of inward leakage by chemicals or particles
  • Offers Til class 2 in Particulate Radioactive Contamination EN1073-2, meaning it has very good particle hold out as well as liquid spray resistance
  • Made with a non-microporous PE laminate for chemical protection with permeation-tested resistance against certain chemicals
  • Knitted cuffs, elasticated waist and ankles make the wearer more comfortable and increase freedom of movement, while the ultra-low linting material reduces environmental contamination
  • Anti-static treatment meeting EN 1149, which reduces static discharge and risk of spark and ignition
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Coveralls
Size Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type 2-way Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Bloodborne Pathogens
No. of Pockets None
For Use With (Application) Controlled Environments
Cleanroom Class Class 10-100,000 (ISO 4-8)