Learn More
Melford Protease Inhibitor Cocktail VI, Plant Cell
Description
Mixture of six protease inhibitors for use with plant cell extracts.
Mixture of six protease inhibitors for use with plant cell extracts. Supplied in 1ml DMSO. Will inhibit proteases extracted from 30g plant tissue.
Contains:
AEBSF
Bestatin
E-64
Leupeptin
Pepstatin A
Phenanthroline
Specificity:
Serine Proteases
Aminopeptidases
Cysteine
Proteases
Aspartic Proteases and Metalloproteases
Specifications
Specifications
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.