Learn More
MedChemExpress Protease Inhibitor Cocktail, mini-Tablet
Specifications
Specifications
| Product Type | Kits |
| Content And Storage | Protease Inhibitor Cocktail (EDTA free, mini-Tablet) can be stored at 4°C for 2 years. |
| Shipping Condition | Room temperature |
| Quantity | 50 Tablets |
| Target | VD/VDR |