Learn More
MedChemExpress Protease-Activated Receptor-4
Specifications
Specifications
| Purity | 98.14% |
| CAS | 245443-52-1 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| Molecular Weight (g/mol) | 666.77 g/mol |
| Product Type | Peptides |
| Quantity | 10 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 50 mg/mL (74.99 mM∣Need ultrasonic) |