missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PROSER1 Polyclonal Antibody

Product Code. 15920705
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15920705 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15920705 Supplier Invitrogen™ Supplier No. PA572025
  • ONLINE EXCLUSIVE PRICE
    530.00 EUR
    514.10 EUR / 400µL
    Save 15.90 EUR
    3% Off
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PROSER1 Polyclonal Antibody for Western Blot

This gene encodes a conserved protein containing proline and serine rich regions. These regions may be important in protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PROSER1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PROSER1
Gene Accession No. Q5PRE5, Q86XN7
Gene Alias 2810046L04Rik; 9330161F11; C13orf23; KIAA2032; proline and serine rich 1; proline and serine-rich protein 1; PROSER1
Gene Symbols PROSER1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 128-156 amino acids from the N-terminal region of human PROSER1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212127, 80209
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.