missing translation for 'onlineSavingsMsg'
Learn More

Propyzamide, TRC

Product Code. 30138027
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138027 1 g 1g
17923695 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138027 Supplier Toronto Research Chemicals Supplier No. TRCP8384001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23950-58-5
Molecular Formula C12 H11 Cl2 N O
Molecular Weight (g/mol) 256.13
Synonym Benzamide, 3,5-dichloro-N-(1,1-dimethyl-2-propyn-1-yl)-, Benzamide, 3,5-dichloro-N-(1,1-dimethyl-2-propynyl)- (8CI,9CI), 3,5-Dichloro-N-(1,1-dimethyl-2-propyn-1-yl)benzamide, 3,5-Dichloro-N-(1,1-dimethyl-2-propynyl)benzamide, 3,5-Dichloro-N-(1,1-dimethylpropynyl)benzamide, Kerb, Kerb 50W, N-(1,1-Dimethylpropynyl)-3,5-dichlorobenzamide, Pronamide, Propyzamide, RH 315
IUPAC Name 3,5-dichloro-N-(2-methylbut-3-yn-2-yl)benzamide
SMILES CC(C)(NC(=O)c1cc(Cl)cc(Cl)c1)C#C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H11Cl2NO/c1-4-12(2,3)15-11(16)8-5-9(13)7-10(14)6-8/h1,5-7H,2-3H3,(H,15,16)
Formula Weight 255.0218
Chemical Name or Material Propyzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.