missing translation for 'onlineSavingsMsg'
Learn More

Propyzamide, Dr. Ehrenstorfer

Product Code. 30321586
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321586 Supplier Dr. Ehrenstorfer Supplier No. DREC16540000100MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propyzamide
CAS 23950-58-5
Analyte or Component Names Propyzamide
Shipping Condition +20°C
Molecular Formula C12 H11 Cl2 N O
Certifications/Compliance ISO 17034
Synonym Benzamide, 3,5-dichloro-N-(1,1-dimethyl-2-propyn-1-yl)-, Benzamide, 3,5-dichloro-N-(1,1-dimethyl-2-propynyl)- (8CI,9CI), 3,5-Dichloro-N-(1,1-dimethyl-2-propyn-1-yl)benzamide, 3,5-Dichloro-N-(1,1-dimethyl-2-propynyl)benzamide, 3,5-Dichloro-N-(1,1-dimethylpropynyl)benzamide, Kerb, Kerb 50W, N-(1,1-Dimethylpropynyl)-3,5-dichlorobenzamide, Pronamide, Propyzamide, RH 315
InChI Formula InChI=1S/C12H11Cl2NO/c1-4-12(2,3)15-11(16)8-5-9(13)7-10(14)6-8/h1,5-7H,2-3H3,(H,15,16)
SMILES CC(C)(NC(=O)c1cc(Cl)cc(Cl)c1)C#C
IUPAC Name 3,5-dichloro-N-(2-methylbut-3-yn-2-yl)benzamide
Molecular Weight (g/mol) 256.13
Quantity 100 mg
Formula Weight 255.0218 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.