missing translation for 'onlineSavingsMsg'
Learn More

Propionic Acid, Sigma-Aldrich™

Product Code. 15858340
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15858340 2.5 L 2.5 L
15816250 100 mL 100mL
15826250 500 mL 500mL
15848340 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15858340 Supplier Sigma Aldrich Supplier No. 8006052500
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-09-4

Specifications

Melting Point -20°C
Density 0.993 g/cm3 at 20°C
Boiling Point 140.7°C to 141.4°C/1013 hPa
MDL Number MFCD00002756
Quantity 2.5 L
Synonym Propionic acid, Methyl acetic acid, Propionic acid
Molecular Weight (g/mol) 74.08
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.