Learn More
Propargyl Benzyl Ether, TRC
Product Code. 30423259
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423259 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30423259 Supplier Toronto Research Chemicals Supplier No. TRCP76256025G
 Restricted Item
Estimated Shipment: 01-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Propargyl Benzyl Ether
CAS 4039-82-1
Molecular Formula C10H10O
Quantity 25 g
Synonym (((Propargyl)oxy)methyl)benzene, 1-(Benzyloxymethyl)ethyne, 1-Benzyloxy-2-propyne, 2-Propynyl benzyl Ether, 3-(Benzyloxy)-1-propyne, 3-(Benzyloxy)propyne, Benzyl 2-propynyl Ether, Benzyl Propargyl Ether, O-Benzyl Propargyl Alcohol, [(2-Propynyloxy)methyl]benzene
InChI Formula InChI=1S/C10H10O/c1-2-8-11-9-10-6-4-3-5-7-10/h1,3-7H,8-9H2
SMILES C#CCOCc1ccccc1
IUPAC Name prop-2-ynoxymethylbenzene
Molecular Weight (g/mol) 146.19
Formula Weight 146.0732