Learn More
Propane-1-sulfonamide, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 24243-71-8 |
| Molecular Formula | C3H9NO2S |
| MDL Number | MFCD03550611 |
| Quantity | 25 g |
| SMILES | CCCS(=O)(N)=O |
| Molecular Weight (g/mol) | 123.17 |
| Percent Purity | 97% |
| Chemical Name or Material | Propane-1-sulfonamide |