Learn More
Propamocarb, TRC
Specifications
Specifications
| Chemical Name or Material | Propamocarb free base |
| CAS | 24579-73-5 |
| Purity Grade Notes | GC |
| Molecular Formula | C9 H20 N2 O2 |
| Quantity | 1 g |
| Synonym | Propamocarb free base, Carbamic acid','N-[3-(dimethylamino)propyl]-','propyl ester, Carbamic acid','[3-(dimethylamino)propyl]-','propyl ester (8CI,9CI), N-(γ-Dimethylaminopropyl)carbamic acid propyl ester, Nor-Am 39744, Plantacur, Plantacur E, Propamocarb, SN 39744 |
| InChI Formula | InChI=1S/C9H20N2O2/c1-4-8-13-9(12)10-6-5-7-11(2)3/h4-8H2,1-3H3,(H,10,12) |
| SMILES | CCCOC(=O)NCCCN(C)C |
| IUPAC Name | propyl N-[3-(dimethylamino)propyl]carbamate |
| Molecular Weight (g/mol) | 188.27 |
| Show More |