Learn More
Propachlor-ethane sulfonic acid (ESA) sodium 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30315753
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315753 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30315753 Supplier Dr. Ehrenstorfer Supplier No. DREA16380210AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Propachlor ethane sulfonic acid (ESA) sodium salt
CAS 947601-88-9
Analyte or Component Names Propachlor ethane sulfonic acid (ESA) sodium salt
Shipping Condition +20°C
Molecular Formula C11 H14 N O4 S . Na
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Sodium 2-[(1-methylethyl)phenylamino]-2-oxoethanesulfonate (1:1)
InChI Formula InChI=1S/C11H15NO4S.Na/c1-9(2)12(10-6-4-3-5-7-10)11(13)8-17(14,15)16;/h3-7,9H,8H2,1-2H3,(H,14,15,16);/q;+1/p-1
SMILES [Na+].CC(C)N(C(=O)CS(=O)(=O)[O-])c1ccccc1
IUPAC Name sodium;2-oxo-2-(N-propan-2-ylanilino)ethanesulfonate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 279.29
Quantity 1 mL
Formula Weight 279.0541 g/mol