Learn More
Serva Electrophoresis™ Pronase E from Streptomyces griseus min. 6.0 DMC-U/mg
Product Code. 11478622
Encompass_Preferred
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. 11478622
Brand: Serva Electrophoresis™ 3363502
Only null left
Specifications
Quantity 1 g
compliance-icons compliance-icons
Product Identifier
  • Pronase E from Streptomyces griseus min. 6 DMC-U/mg
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .