missing translation for 'onlineSavingsMsg'
Learn More

Progabide, TRC

Product Code. 30420946
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420946 1 g -
30414004 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30420946 Supplier Toronto Research Chemicals Supplier No. TRCP7553501G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62666-20-0
Molecular Formula C17 H16 Cl F N2 O2
Molecular Weight (g/mol) 334.77
Synonym Butanamide, 4-[[(4-chlorophenyl)(5-fluoro-2-hydroxyphenyl)methylene]amino]-, 4-[[(4-Chlorophenyl)(5-fluoro-2-hydroxyphenyl)methylene]amino]butanamide, Gabren, Gabrene, Halogabide, Progabide, SL 76-002, SL 76002
IUPAC Name 4-[(Z)-[(4-chlorophenyl)-(5-fluoro-2-hydroxyphenyl)methylidene]amino]butanamide
SMILES NC(=O)CCC\N=C(\c1ccc(Cl)cc1)/c2cc(F)ccc2O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H16ClFN2O2/c18-12-5-3-11(4-6-12)17(21-9-1-2-16(20)23)14-10-13(19)7-8-15(14)22/h3-8,10,22H,1-2,9H2,(H2,20,23)/b21-17-
Formula Weight 334.0884
Chemical Name or Material Progabide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.