missing translation for 'onlineSavingsMsg'
Learn More

Procyclidine hydrochloride, MedChemExpress

Product Code. 16438776
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438776 Supplier Medchem Express Supplier No. HYB1487
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Procyclidine (Tricyclamol, (±)-Procyclidine) hydrochloride , an anticholinergic agent, is a muscarinic receptor antagonist that also has the properties of an N-methyl-D-aspartate (NMDA) antagonist. Procyclidine hydrochloride can be used in studies of Parkinson's disease and related psychiatric disorders such as Soman-induced epilepsy.

Procyclidine (Tricyclamol, (±)-Procyclidine) hydrochloride , an anticholinergic agent, is a muscarinic receptor antagonist that also has the properties of an N-methyl-D-aspartate (NMDA) antagonist. Procyclidine hydrochloride can be used in studies of Parkinson's disease and related psychiatric disorders such as Soman-induced epilepsy.

Specifications

Chemical Name or Material Procyclidine hydrochloride
CAS 1508-76-5
Color White
Purity Grade Notes Research
Molecular Formula C19H30ClNO
Quantity 100 mg
Synonym Tricyclamol hydrochloride(±)-Procyclidine hydrochloride
Solubility Information DMSO : 10.66 mg/mL (32.91 mM; Need ultrasonic and warming) ∣H2O : 5.56 mg/mL (17.17 mM; ultrasonic and warming and heat to 60°C)
SMILES OC(C1=CC=CC=C1)(C2CCCCC2)CCN3CCCC3.[H]Cl
Molecular Weight (g/mol) 323.9
Formula Weight 323.9
Percent Purity 98.8%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.