Learn More
Prochlorperazine, TRC
Product Code. 30136995
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136995 50 mg 50mg
30135423 25 mg 25mg
17974825 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136995 Supplier Toronto Research Chemicals Supplier No. TRCP75579050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 58-38-8
Molecular Formula C20 H24 Cl N3 S
Molecular Weight (g/mol) 373.94
Synonym 10H-Phenothiazine, 2-chloro-10-[3-(4-methyl-1-piperazinyl)propyl]-, Phenothiazine, 2-chloro-10-[3-(4-methyl-1-piperazinyl)propyl]- (7CI,8CI), 2-Chloro-10-(3-[1-methyl-4-piperazinyl]propyl)phenothiazine, 2-Chloro-10-[3-(4-methyl-1-piperazinyl)propyl]-10H-phenothiazine, 2-Chloro-10-[3-(4-methyl-1-piperazinyl)propyl]phenothiazine, 3-Chloro-10-[3-(1-methyl-4-piperazinyl)propyl]phenothiazine, 6140RP, Bayer A 173, Capazine, Chlormeprazine, Chlorperazine, DDD 66604, Emelent, Kronocin, Meterazin, Meterazine, N-[γ-(4'-Methylpiperazinyl-1')propyl]-3-chlorophenothiazine, NSC 17478, NSC 665801, Nipodal, Proazine, Prochlorpemazine, Prochlorperazin, Prochlorperazine, Prochlorpromazine, RP 6140, SKF 4657, Temetid
IUPAC Name 2-chloro-10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine
SMILES CN1CCN(CCCN2c3ccccc3Sc4ccc(Cl)cc24)CC1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C20H24ClN3S/c1-22-11-13-23(14-12-22)9-4-10-24-17-5-2-3-6-19(17)25-20-8-7-16(21)15-18(20)24/h2-3,5-8,15H,4,9-14H2,1H3
Formula Weight 373.1379
Percent Purity >95%
Chemical Name or Material Prochlorperazine
Grade HPLC