Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Prochloraz |
| CAS | 67747-09-5 |
| CAS Max % | ≤ 100% |
| Linear Formula | C15H16Cl3N3O2 |
| Beilstein | 8155344 |
| Color | White |
| Melting Point | 46.3°C to 50.3°C |
| Synonym | N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]-1H-imidazole-1-carboxamide |
| Molecular Weight (g/mol) | 376.7 g/mol |
| Quantity | 100 mg |