missing translation for 'onlineSavingsMsg'
Learn More

Procainamide hydrochloride, MedChemExpress

Product Code. 16448026
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448026 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16448026 Supplier Medchem Express Supplier No. HYA0084
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Procainamide hydrochloride is an anti-arrhythmic agent and is used to treat cardiac arrhythmia; induces rapid block of the batrachotoxin(BTX)-activated sodium channels of the heart muscle and acts as antagonist to long gating closures.

Procainamide hydrochloride is an anti-arrhythmic agent and is used to treat cardiac arrhythmia

Specifications

Chemical Name or Material Procainamide hydrochloride
Physical Form Solid
CAS 614-39-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H22ClN3O
For Use With (Application) Cancer-programmed cell death
Solubility Information H2O : ≥ 50 mg/mL (183.97 mM) ∣DMSO : 50 mg/mL (183.97 mM; Need ultrasonic)
SMILES O=C(NCCN(CC)CC)C1=CC=C(N)C=C1.[H]Cl
Molecular Weight (g/mol) 271.79
Formula Weight 271.79
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.