Learn More
Probucol, Mikromol™
Product Code. 30052043
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052043 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052043 Supplier Mikromol Supplier No. MM1068.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Probucol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Probucol
CAS 23288-49-5
Analyte or Component Names Probucol
Shipping Condition Room Temperature
Molecular Formula C31 H48 O2 S2
Certifications/Compliance ISO 17025
Synonym Phenol, 4,4'-[(1-methylethylidene)bis(thio)]bis[2,6-bis(1,1-dimethylethyl)-, Acetone, bis(3,5-di-tert-butyl-4-hydroxyphenyl) mercaptole (8CI), Phenol, 4,4'-(isopropylidenedithio)bis[2,6-di-tert-butyl- (8CI), 4,4'-(Isopropylidenedithio)bis[2,6-di-tert-butylphenol], Biphenabid, Bisbid, Bisphenabid, DH 581, Lipomal, Lorelco, Lurselle, NSC 652160, NSC 86225, Panavir, Phenbutol, Probucol, Sinlestal
InChI Formula InChI=1S/C31H48O2S2/c1-27(2,3)21-15-19(16-22(25(21)32)28(4,5)6)34-31(13,14)35-20-17-23(29(7,8)9)26(33)24(18-20)30(10,11)12/h15-18,32-33H,1-14H3
SMILES CC(C)(C)c1cc(SC(C)(C)Sc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C
IUPAC Name 2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol
Molecular Weight (g/mol) 516.84
Quantity 250 mg
Formula Weight 516.3096 g/mol