missing translation for 'onlineSavingsMsg'
Learn More

Probucol, 99.7%, MP Biomedicals™

Product Code. 11453752 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453752 50 g 50g
11433752 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11453752 Supplier MP Biomedicals Supplier No. 0215637150
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 23288-49-5
Molecular Formula C31H48O2S2
Molecular Weight (g/mol) 516.843
InChI Key FYPMFJGVHOHGLL-UHFFFAOYSA-N
Synonym probucol, lorelco, biphenabid, bisphenabid, lurselle, bisbid, lesterol, lursell, panavir, probucolum
PubChem CID 4912
ChEBI CHEBI:8427
IUPAC Name 2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)SC(C)(C)SC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C

Specifications

Color White
Quantity 50 g
Formula Weight 516.8
Percent Purity 0.997
Physical Form Powder
Chemical Name or Material Probucol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.