Learn More
Probucol, 99.7%, MP Biomedicals™
Product Code. 11433752 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433752 5 g 5g
11453752 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11433752 Supplier MP Biomedicals Supplier No. 0215637105
Chemical Identifiers
CAS 23288-49-5
Molecular Formula C31H48O2S2
Molecular Weight (g/mol) 516.843
InChI Key FYPMFJGVHOHGLL-UHFFFAOYSA-N
Synonym probucol, lorelco, biphenabid, bisphenabid, lurselle, bisbid, lesterol, lursell, panavir, probucolum
PubChem CID 4912
ChEBI CHEBI:8427
IUPAC Name 2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)SC(C)(C)SC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C
Specifications
Color White
Quantity 5 g
Formula Weight 516.8
Percent Purity 0.997
Physical Form Powder
Chemical Name or Material Probucol