Learn More
Epredia™ PrintMate™ AS Cassette Printer Accessories
Product Code. 12973745 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Cassette Hopper
Collection System
Collection Tray
Computer
Hot Foil Tape
Unit Size:
Each
Pack of 4
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12973745 Collection System Each
15589535 Collection Tray Pack of 4
12628206 Hot Foil Tape Pack of 5
12963815 Computer Each
12648206 Cassette Hopper Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12973745 Supplier Epredia™ Supplier No. A79510014
  • ONLINE EXCLUSIVE PRICE
    18180.00 EUR
    14191.95 EUR / Pack of 1
    Save 3988.05 EUR
    21% Off
Estimated Shipment: 16-11-2026
Maintain the Epredia™ PrintMate™ Cassette Printer with PrintMate™ Accessories.
Accessories Include:
  • Collection system collection trays (box of 4)
  • Hopper
  • Hot foil tape, box of 5 (roll of tape prints approx. 20,000 cassettes)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Collection System
Description Collection system
For Use With (Equipment) PrintMate AS Cassette Printer, PrintMate Cassette Printer