Learn More
JULABO PRESTO Process Systems
GREENER_CHOICE
Product Code. 17714869
Change view
Click to view available options
Model:
A38
A41
A70
W41
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
17714869 A38 Each
17724869 A41 Each
17734869 A70 Each
17744869 W41 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17714869 Supplier JULABO Supplier No. 9420381.N1
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
The PRESTO systems are designed for precise temperature control as well as rapid temperature changes, making them ideal for reactor vessels, material stress tests, or temperature simulations.
  • Wide working temperature ranges without changing fluids
  • Ethernet interface
  • SD-Card slot
  • pace optimized design creates more room directly next to the units
  • Connections for USB, Ethernet, RS232, and Alarm output
Specifications
Temperature Range (Metric) -30°C to +250°C
Dimensions (L x W x H) 29.5 x 13 x 26.4 in.
Model A38
Type Process System