Description
- With DIN thread, GL 45
- Thermal shock resistance of 160 K
- Accuracy scale of ± 5%
- Applications include the pharmaceutical industry, handling of aggressive media, sterilization processes (hot air and dry) and depyrogenation
- Without screw cap
- Provided in packs of 10
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Color | Clear |
| For Use With (Application) | Holding, measuring |
| Material | Borosilicate Glass |
| Max. Temperature (Metric) | 500°C |
| Quantity | 10/pk |
| Thread Size (Metric) | GL 45 |
| Closure Included | No |
| Graduated | No |
| Height (Metric) | 100 mm |
| Show More |