Learn More
Uvex Preformed Mask silv-Air p 7110 FFP1
Product Code. 17572945 Shop All Uvex Products
Change view
Click to view available options
Filter Type: FFP1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17572945 Filter Type: FFP1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17572945 Supplier Uvex Supplier No. 8707110
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The gray coloring on the outside of the silv-Air p masks prevents them from looking dirty quickly and having to be disposed of prematurely by the user. By contrast, the inside is of course pure white. In addition and with all products in this family, the 3D nose geometry ensures an optimum fit, even with different face shapes.
General Features
  • Particle-filtering preformed mask with exhalation valve to reduce exhalation resistance and heat and moisture build-up inside the mask
  • Metal-free processing
  • Soft material edges for increased wearer comfort
  • Seamless headband for a comfortable, secure fit
  • Color: silver-gray
Protection Features
  • FFP1 respirator certified in accordance with EN 149:2001+A1:2009
  • D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
  • Not reusable
Comfort Features
  • Soft material edges for increased wearer comfort
  • Seamless headband for a comfortable, secure fit
  • Exhalation valve to reduce breathing resistance and minimise heat and moisture build-up inside the mask
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149 (D marked)
Design Type Cup/Molded
Disposable Single-use
Material Polypropylene, Polyester
Product Type Preformed Mask
Size Standard
With Exhalation Valve Yes
Filter Efficiency FFP1
Sterility Non-sterile