Learn More
Uvex Preformed Mask silv-Air e 7233 FFP2
Product Code. 17522955 Shop All Uvex Products
Change view
Click to view available options
Filter Type: FFP2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17522955 Filter Type: FFP2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17522955 Supplier Uvex Supplier No. 8707233
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
All masks of the silv-Air e family combine the unique technologies of optimum wearer comfort with extremely low breathing resistance. The products are therefore suitable for heavy work and for being worn for long periods of time. The increased size of the filter surface even enables the high-performance models to be worn for several days.
General Features
  • Particle-filtering preformed mask with exhalation valve and inhalation valves
  • Color: white
Protection Features
  • Certified FFP2 face mask to EN 149:2001+A1:2009
  • D label (dolomite dust test passed): maximum breathability even with really high dust levels
  • R label (reusable): tested for re-usability and performance, including in the second shift
Comfort Features
  • High-performance climazone system with three compartments for a comfortable, cool climate inside the mask
  • Flexible textile headband with neck clip for individual adjustment
  • Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
  • Two extra filters with inhalation valves for lower breathing resistance and increased comfort due to easier air exchange
  • Very soft, all-around sealing lip for optimum wearer comfort and secure fit
  • Combination of 3D nose area and sealing lip ensures ideal fit for almost any nose shape
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149 (R marked, D marked)
Product Type Preformed Mask
With Exhalation Valve Yes
Filter Efficiency FFP2
Includes 360° exhalation valve, 3D nose section, circumferential comfort sealing lip