Learn More
MilliporeSigma™ Precoated HPTLC Glass Plates
Description
- Average particle size is 5 μm with a 4-7 μm distribution
- Untreated, chemically modified, and reversed-phase silica gel with a pore size of 60 Å
- For all common gel separations, including weak to strong polar analytes, for applications requiring charring procedures
Certifications
ISO 9001:2000
Specifications
Specifications
| Backing Material | Glass |
| Material | Silica 60 |
| Particle Size | 5 to 6 μm |
| Pore Size | 60 Å |
| Quantity | 50/Pk. |
| Type | Silica HPTLC Plate |
| Fluorescent Indicator | F254 |
| For Use With (Application) | Thin layer chromatography (TLC), in-process control in drug synthesis, identity and stability testing of drugs, quality control of pharmaceuticals, food and environmental samples, residue analysis in food and environmental samples |
| Sorbent | Silica Gel 60 Premium |
| Thickness (Metric) | 200 μm |
| Show More |