Learn More
Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil
Product Code. 12612810 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (English) Inner:
0.003 in.
0.005 in.
0.007 in.
0.010 in.
0.020 in.
0.030 in.
0.040 in.
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Unit Size
12612810 0.007 in. Each
10428114 0.003 in. Each
10421385 0.005 in. Each
11577691 0.007 in. Each
10013984 0.010 in. Each
10659875 0.020 in. Each
10197364 0.030 in. Each
10260535 0.040 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 12612810
  • ONLINE EXCLUSIVE PRICE
    42.80 EUR
    41.93 EUR / Pack of 1
    Save 0.87 EUR
    2% Off
View all promotions
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil is easily cut to the desired length.
Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil is easily cut to the desired length.
  • Broad chemical compatibility
  • Biocompatible
  • May be used with standard fingertight fittings
  • Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
TRUSTED_SUSTAINABILITY
Specifications
For Use With HPLC
Type HPLC Tubing
For Use With (Application) HPLC
Diameter (English) Inner 0.007 in.
Diameter (Metric) Inner 0.18 mm
Overall Length (English) 5 ft.
Overall Length (Metric) 1.524 m
Material PEEK