Learn More
Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil
Product Code. 10260535 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (English) Inner:
0.003 in.
0.005 in.
0.007 in.
0.010 in.
0.020 in.
0.030 in.
0.040 in.
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Unit Size
10260535 0.040 in. Each
10428114 0.003 in. Each
10421385 0.005 in. Each
12612810 0.007 in. Each
11577691 0.007 in. Each
10013984 0.010 in. Each
10659875 0.020 in. Each
10197364 0.030 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10260535 Supplier Thermo Scientific™ Supplier No. 37040
Estimated Shipment: 22-12-2026
Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil is easily cut to the desired length.
Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil is easily cut to the desired length.
  • Broad chemical compatibility
  • Biocompatible
  • May be used with standard fingertight fittings
  • Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
TRUSTED_SUSTAINABILITY
Specifications
For Use With HPLC
Type HPLC Tubing
Description 0.040 in. ID; 5 ft. length; Natural
For Use With (Application) HPLC
Diameter (English) Inner 0.040 in.
Diameter (Metric) Inner 1.016 mm
Overall Length (English) 5 ft.
Overall Length (Metric) 1.524 m
Material PEEK