Learn More
Grant Instruments™ PQ Documentation
Product Code. 15211216
Change view
Click to view available options
For Use With (Equipment):
All BTD Models
All Grant XUB Digital Ultrasonic Baths
All Grant XUBA Analogue Ultrasonic Baths
All JB Aqua Plus Models (JBN and JBA)
All QBD Models
All QBH Models
All SUB Aqua Pro Models (SAP)
Grant CS200G Stirred Thermostatic Bath/Circulator
Grant CVP-2 PCR Plate Centrifuge
Grant CXG Stirred Thermostatic Bath/Circulator
Grant PHMP-100 Thermoshaker for Microplates
Grant R4 Refrigerated Baths/Circulators
Grant T100 Stirred Thermostatic Bath/Circulator
Grant TC120 Stirred Thermostatic Bath/Circulator
Grant TS-DW Deep Well Plate Thermoshaker
Grant TX150 Stirred Thermostatic Bath/Circulator
Grant TXF200 Stirred Thermostatic Bath/Circulator
Unit Size:
Each
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15211216 All QBH Models Each
15251216 Grant R4 Refrigerated Baths/Circulators Each
15201226 Grant TC120 Stirred Thermostatic Bath/Circulator Each
15261216 All SUB Aqua Pro Models (SAP) Each
15211226 All Grant XUB Digital Ultrasonic Baths Each
15201216 All QBD Models Each
15251206 All BTD Models Each
15221226 All Grant XUBA Analogue Ultrasonic Baths Each
15271206 All JB Aqua Plus Models (JBN and JBA) Each
15152615 Grant TX150 Stirred Thermostatic Bath/Circulator Each
15122625 Grant CS200G Stirred Thermostatic Bath/Circulator Each
15192615 Grant CXG Stirred Thermostatic Bath/Circulator Each
15132615 Grant T100 Stirred Thermostatic Bath/Circulator Each
15172615 Grant TXF200 Stirred Thermostatic Bath/Circulator Each
15420484 Grant CVP-2 PCR Plate Centrifuge Each
15450484 Grant PHMP-100 Thermoshaker for Microplates Each
15490474 Grant TS-DW Deep Well Plate Thermoshaker Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 15211216 Supplier Grant Instruments™ Supplier No. PQ QBH
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
Offers equipment user greater comparability and verification with their instruments. Grant Instruments™ PQ Documentation simplifies installation and maintenance procedures, reduces common user errors, and builds a clear record of an instrument's performance throughout its life.
Performance Qualification (PQ): Instrument is continuing to meet its specification
  • Each documentation set is specific to individual serial numbered equipment and is provided in an easy to use ring bound format.
  • PQ documentation can be ordered at any time.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) All QBH Models
Product Type PQ Documentation