missing translation for 'onlineSavingsMsg'
Learn More

PPP2R1A Antibody, Novus Biologicals™

Product Code. 30228048 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30228048 100 μL 100µL
30231168 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30228048 Supplier Novus Biologicals Supplier No. NBP338186100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPP2R1A Polyclonal antibody specifically detects PPP2R1A in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R1A
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:10 - 1:100
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias Medium tumor antigen-associated 61 kDa protein, MGC786, PP2A subunit A isoform PR65-alpha, PP2A subunit A isoform R1-alpha, PP2A-Aalpha, PP2AAALPHA, PR65A, protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alphaisoform, protein phosphatase 2 (formerly 2A), regulatory subunit A, alpha isoform, protein phosphatase 2, regulatory subunit A, alpha, serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alphaisoform, serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alphaisoform
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human PPP2R1A (NP_055040.2).,, Sequence:, MAAADGDDSLYPIAVLIDELRNEDVQLRLNSIKKLSTIALALGVERTRSELLPFLTDTIYDEDEVLLALAEQLGTFTTLVGGPEYVHCLLPPLESLATVEETVVRDKAVESLRAISHEHSPSDLEAHFVPLVKRLAGGDWFTSRTSACGLFSVCYPRVSSAVKAELRQYFRNLCSDDTPMVRRAAASKLGEFAKVLELDNVKSEIIPMFSNLASDEQDSVRLLAVEACVNIAQLLPQEDLEALVMPTLRQAAEDKSWRVRYMVADKFTEL
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5518
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.