Learn More
Bel-Art™ Scienceware™ Poxy Coated Wire Framed Bag Holder
Product Code. 10633305
Change view
Click to view available options
Color:
Orange
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10633305 Orange Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10633305 Supplier Bel-Art™ Scienceware™ Supplier No. H131931000
  • ONLINE EXCLUSIVE PRICE
    30.25 EUR
    24.76 EUR / Pack of 1
    Save 5.49 EUR
    18% Off
Estimated Shipment: 20-11-2026
Collects pipette tips, microcentrifuge tubes. Bel Art™ Poxy Coated Wire Framed Bag Holder is economical and convenient.
  • Rectangular steel wire frame holds a polyethylene bag 22 x 28cm (8½ x 11 in.); coated in easy to clean, durable, Orange color epoxy
  • Snug fitting polypropylene plastic cover helps hold the bag securely in place
  • Reusable; can be cleaned by hand or in automated dish washers; autoclavable.
  • Cover provides compliance with 29 CFR 1910.1030(d)(2)(xiii)(A), which requires a cover on any container used to store, transport, or ship biohazardous wastes
  • Can be used with Bench Top Biohazard Bags (F13166-0000); can also be used with Clavies Autoclavable Bags (H13182-0812)
Specifications
Autoclavable Autoclavable
Color Orange
Disposable Reusable
For Use With (Equipment) For bench top biohazard bags
Material Epoxy-coated Wire
Width (English) 8.5 in.
Product Type Bag Holder
Width (Metric) 22 cm