Learn More
Ansell™ Powerflex™ 80-409 (ex Nitrotough™ N1700 FD Insulator™)
Product Code. 15814992 Shop All Ansell Products
Change view
Click to view available options
Size:
8
9
10
Unit Size:
6 pairs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15814992 10 6 pairs
15894982 8 6 pairs
15804992 9 6 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15814992 Supplier Ansell™ Supplier No. 80409100
Estimated Shipment: 07-01-2027
High flexibility and dexterity for an optimum comfort in use
  • Seamless nylon liner, gauge 13.
  • Rough nitrile coating.
  • Fully dipped.
  • Fine and stretchable stitches for excellent comfort.
  • Elastic knitwrist ensures snug fit.
  • Excellent abrasion resistance.
  • Covered in a layer resistant to polluted pieces being handled.
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Certifications/Compliance EN388:2016/4121C, EN511/020
Classification Category II
Cleaning Procedure Washing temp 40° C
Coated Area fully coated
Cuff Material Nitrile
Cut Style Number
Description Glove for all type of work various handling works in difficult thermal conditions
Coating Material Nitrile Rubber, Polyurethane
Color Blue
Cut Resistance Rating Level 1
Disposable Reusable
Exterior Finish Fully Textured
Grade Industrial grade
Hand Specific
Latex Free Yes
Material Polyurethane
Powdered Powder Free
Product Type Mechanical Protection Gloves (Medium-duty)
Size 10
Sterility Non-sterile
Temperature (English) Max. 5°F
Temperature (Metric) Max -15°C
Thickness (Metric) 2.65 mm
Finger Style Fully fingered
For Use With (Application) Mechanical protection
Glove Cuff Color White
Glove Cuff Style Automatic Knit Liner
Liner Material Acrylic, Polyester
Lining Knit Lining
Model 80-409
Packaging Type 6 pair in a bag, 12 bags in a carton
Puncture Rating Level 1
Quantity 72 pair
Series Powerflex series
Tear Rating 2
Dimensions (L x W) 275 x 129 mm
Length (English) 275 mm
Width (Metric) 129 mm
Weight (Metric) 72 g