Learn More
Invitrogen™ Power Blotter System
Product Code. 15806162
Change view
Click to view available options
Quantity:
1 system
missing translation for 'unitSize':
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity missing translation for 'unitSize'
15806162 1 system Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15806162 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' PB0012
Protein Biology Essentials
Use promo code 27946
Ver todas las promociones
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones
Includes
Invitrogen Power Blotter Cassette, Invitrogen Power Power Station
Get rapid and efficient semi-dry protein transfer with the Invitrogen™ Power Blotter.
The Invitrogen™ Power Blotter System is specifically designed for rapid semi-dry transfer of proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes in 5 to 12 minutes. The Power Blotter includes an integrated power supply optimized to enable consistent and high-efficiency protein transfer.
The Power Blotter Cassette enables the simultaneous transfer of gels. The Power Blotter Station has an easy-to-use color LCD touchscreen interface with pre-programmed transfer methods for different numbers and sizes of gels and ranges of molecular weight proteins. The easy-touch programming feature allows custom transfer settings to be quickly created, saved, and run.
The Power Blotter achieves highly efficient transfer in 7 minutes when used with Power Blotter Select Transfer Stacks or 10 minutes with the Power Blotter 1-Step Transfer Buffer. The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional Towbin buffer.
Features:
  • Efficient—high transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wet (tank) transfer methods
  • Fast—5 to 12 minute transfer when used with Power Blotter Select Transfer Stacks or Power Blotter 1-Step Transfer Buffer
  • Integrated power supply—seamless operation between control unit and cassette; no need for external power supply
  • Easy-touch programming—convenient pre-programmed transfer methods or create, save, and run customized transfer methods
  • Flexible—simultaneous transfer
  • Versatile—compatible with Power Blotter Select Transfer Stacks, Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods
TRUSTED_SUSTAINABILITY
Especificaciones
Content And Storage • Power Blotter Station
• Power Blotter Cassette
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Mode of Transfer Semi-Dry
Product Line Power Blotter
Throughput 1 Midi or 2 Mini Gels
Type Blotter System
Quantity 1 system
Shipping Condition Room Temperature
For Research Use Only. Not for use in diagnostic procedures.
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.