missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate, Supelco™

Product Code. 15817330
Change view
Click to view available options
Quantity:
1 kg
50 kg
Unit Size:
1kg
50kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15817330 50 kg 50kg
15807330 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15817330 Supplier Supelco Supplier No. 1051249050
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EMPLURA™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-20-0

Specifications

Boiling Point >400°C/1013 hPa (decomposition)
Melting Point 177°C
Quantity 50 kg
MDL Number MFCD00011413
Synonym Potassium thiocyanate, Potassium sulfocyanide, Potassium thiocyanate
Molecular Weight (g/mol) 97.18
Grade Research
Density 1.91 g/cm3 at 20°C
Chemical Name or Material Potassium Thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.