Learn More
Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 105560-52-9 |
| Molecular Formula | C32H12BF24K |
| MDL Number | MFCD00077934 |
| Quantity | 50 mg |
| SMILES | FC(C1=CC(C(F)(F)F)=CC([B-](C2=CC(C(F)(F)F)=CC(C(F)(F)F)=C2)(C3=CC(C(F)(F)F)=CC(C(F)(F)F)=C3)C4=CC(C(F)(F)F)=CC(C(F)(F)F)=C4)=C1)(F)F.[K+] |
| Molecular Weight (g/mol) | 902.31 |
| Percent Purity | 98% |
| Chemical Name or Material | Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate |