Learn More
Potassium Sulfate, For Analysis, EMSURE™, MilliporeSigma™,
Product Code. 15857330
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15857330 5 kg Plastic Bottle 5kg
15824540 500 g Plastic Bottle 500g
15834540 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15857330 Supplier Supelco Supplier No. 1051535000
 Restricted Item
Estimated Shipment: 26-11-2026
Recommended Storage
+5°C to +30°C.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O
Specifications
Boiling Point 1689°C (1013hPa)
Melting Point 1069°C
Color Colorless
pH 5.5 to 7.5
Physical Form Solid
Quantity 5 kg
Solubility Information 111g/L
Formula Weight 174.26
Percent Purity >99.0%
Grade ACS
Packaging Plastic Bottle
Density 2.66 g/3 (20 °C)