Learn More
Sherwood Scientific™ Potassium Standard Solution
Product Code. 12951851
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12951851 100 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12951851 Supplier Sherwood Scientific™ Supplier No. 00156621
Specifications
Certifications/Compliance NISTM KCl Ref SRM 918
Concentration 1000 ppm
Packaging 6 Pk.
Quantity 100 mL