missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sodium Tartrate Tetrahydrate, Supelco™

Product Code. 15817640
Change view
Click to view available options
Quantity:
50 kg
Unit Size:
50kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15817640 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15817640 Supplier Supelco Supplier No. 1080879050
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS,ISO,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 70°C to 80°C
CAS 6381-59-5
Quantity 50 kg
MDL Number MFCD00150989
Synonym Potassium sodium tartrate tetrahydrate, Sodium potassium tartrate, Tartaric acid potassium sodium salt, Potassium sodium tartrate tetrahydrate
Molecular Weight (g/mol) 282.22
Grade Research
Chemical Name or Material Potassium Sodium Tartrate Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.