missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate dibasic, ACS Reagent, ≥98%, Solstice
Description
Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured
Specifications
Specifications
| CAS | 11-4-7758 |
| Quantity | 25 kg |
| Molecular Formula | HK2O4P |
| InChI Key | ZPWVASYFFYYZEW-UHFFFAOYSA-L |
| SMILES | [K+].[K+].OP([O-])([O-])=O |
| IUPAC Name | dipotassium hydrogen phosphate |
| Molecular Weight (g/mol) | 174.17 |
| Chemical Name or Material | Potassium phosphate dibasic |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?