Learn More
Potassium Permanganate Solution, 0.02mol/L, Chem Lab™
Product Code. 11913183
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11913183 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 11913183 Supplier Chem-Lab Supplier No. CL05.1113.1000
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Specifications
Color Black-Purple
Boiling Point 100°C
Melting Point 0°C
Composition 3.161g KMnO4 / l H2O = 0.1N (±0.0002/20°C)
Density 1g/mL
Solubility Information Water Soluble
Odor Odorless
pH 6 to 8
Quantity 1 L
Physical Form Liquid
compliance-icons
Product Identifier
  • Potassium permanganate 0.02 mol/l
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: potassium permanganate