Learn More
Potassium Orotate, TRC
Product Code. 30138697
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138697 10 g 10g
30134829 5 g 5g
30006843 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138697 Supplier Toronto Research Chemicals Supplier No. TRCP69876010G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24598-73-0
Molecular Formula C5H3N2O4K
Molecular Weight (g/mol) 194.19
IUPAC Name potassium;2,4-dioxo-1H-pyrimidine-6-carboxylate
SMILES C1=C(C(=O)O)N=C(N=C1[O-])O.[K+]
Specifications
Quantity 10 g
InChI Formula InChI=1S/C5H4N2O4.K/c8-3-1-2(4(9)10)6-5(11)7-3;/h1H,(H,9,10)(H2,6,7,8,11);/q;+1/p-1
Formula Weight 193.973
Chemical Name or Material Potassium Orotate