missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, TRC

Product Code. 30008635
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Unit Size:
250g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008635 25 g 25g
30004908 250 g 250g
30003506 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008635 Supplier Toronto Research Chemicals Supplier No. TRCP69509025G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula I K
Molecular Weight (g/mol) 166
Synonym Potassium iodide, Antistrumin, Asmofug E, Ceoiodin, Iod-active, Iodomarin, Iodostin, Jodid, K1-N, Kaiod
IUPAC Name iodopotassium
SMILES [K]I

Specifications

Quantity 25 g
InChI Formula InChI=1S/HI.K/h1H;/q;+1/p-1
Formula Weight 165.8682
Chemical Name or Material Potassium iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.