missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, Supelco™

Product Code. 15844510
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
50 kg
Unit Size:
1kg
2.5kg
250g
500g
50kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15844510 1 kg 1kg
15824510 250 g 250g
15834510 500 g 500g
15854510 2.5 kg 2.5kg
15827310 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15844510 Supplier Supelco Supplier No. 1050431000
Supelco Essentials Are in Stock
Use promo code 27958
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ISO,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0

Specifications

Boiling Point 1325°C/1013 hPa
Melting Point 681°C (lit.)
Quantity 1 kg
Molecular Formula KI
MDL Number MFCD00011405
Molecular Weight (g/mol) 166
Grade Research
Chemical Name or Material Potassium Iodide
compliance-icons compliance-icons
Product Identifier
  • Potassium iodide for analysis EMSURE® ISO,Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.