Learn More
Potassium hexacyanoruthenate(II) hydrate, Ru 23.0% min
Product Code. 11321958
Change view
Click to view available options
Quantity:
0.2 g
2 g
10 g
Unit Size:
0.2g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11321958 2 g 2g
11331958 0.2 g 0.2g
11341958 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11321958 Supplier Thermo Scientific Alfa Aesar Supplier No. 011876.04
 Restricted Item
In Stock
Add to basket
CAS: 339268-21-2 | C6K4N6Ru | 413.57 g/mol
Potassium hexacyanoruthenate(II) hydrate used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 339268-21-2
Molecular Formula C6K4N6Ru
Molecular Weight (g/mol) 413.57
MDL Number MFCD00168064
InChI Key FULSJSKUKUZHLZ-UHFFFAOYSA-N
Synonym potassium hexacyanoruthenate ii hydrate, tetrapotassium hexacyanorutheniumtetrauide hydrate
PubChem CID 71311369
IUPAC Name tetrapotassium;ruthenium(2+);hexacyanide;hydrate
SMILES [K+].[K+].[K+].[K+].N#C[Ru++](C#N)(C#N)(C#N)(C#N)C#N
Specifications
Color White
Physical Form Crystalline
Quantity 2 g
Linear Formula K4Ru(CN)6·xH2O
UN Number UN1588
Solubility Information Soluble in water.
Formula Weight 413.57 (Anhydrous)
Concentration or Composition (by Analyte or Components) Ru 23.0% min.
Chemical Name or Material Potassium hexacyanoruthenate(II) hydrate